N-(3-Acetyl-4-hydroxyphenyl)butyramid 99%

BLDPBD69443-100GEA 50 EUR
BLDPBD69443-100G BLDPBD69443-25G BLDPBD69443-500G
N-(3-Acetyl-4-hydroxyphenyl)butyramid 99%
N-(3-Acetyl-4-hydroxyphenyl)butyramid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₅NO₃
Molekulargewicht: 221,26 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00798556
CAS-Nummer: 40188-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR