D-Phenylalaninol 98%

BLDPBD6967-5GEA 8 EUR
BLDPBD6967-5G BLDPBD6967-500G BLDPBD6967-0.5G BLDPBD6967-1G BLDPBD6967-100G BLDPBD6967-10G BLDPBD6967-0.025G BLDPBD6967-25G
D-Phenylalaninol 98%
D-Phenylalaninol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₁₃NO
Molekulargewicht: 151,2 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064298
CAS-Nummer: 5267-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR