4-Brom-5-chloranthranilsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD70084-1GEA 23 EUR
BLDPBD70084-1G BLDPBD70084-250MG BLDPBD70084-25G BLDPBD70084-10G BLDPBD70084-100G BLDPBD70084-5G
4-Brom-5-chloranthranilsäure 95%
4-Brom-5-chloranthranilsäure

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅BrClNO₂
Molekulargewicht: 250,4800 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10000554
CAS-Nummer: 150812-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR