2-Brom-p-anisidin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD71829-1GEA 10 EUR
BLDPBD71829-1G BLDPBD71829-100G BLDPBD71829-25G BLDPBD71829-5G BLDPBD71829-10G
2-Brom-p-anisidin 98%
2-Brom-p-anisidin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₈BrNO
Molekulargewicht: 202,0485 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09029057
CAS-Nummer: 32338-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR