2-Bromo-4-nitrobenzaldehyde 98%
|
|
BLDPBD71861-25GEA
1264
EUR
BLDPBD71861-25G
BLDPBD71861-5G
BLDPBD71861-100MG
BLDPBD71861-1G
BLDPBD71861-250MG
2-Bromo-4-nitrobenzaldehyde 98%
2-Bromo-4-nitrobenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₄BrNO₃ Molekulargewicht: 230,016 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08753661 CAS-Nummer: 5274-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...