2-Bromo-5-ethoxypyridine 95%

BLDPBD71929-1GEA 48 EUR
BLDPBD71929-1G BLDPBD71929-250MG
2-Bromo-5-ethoxypyridine 95%
2-Bromo-5-ethoxypyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₈BrNO
Molekulargewicht: 202,049 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09260481
CAS-Nummer: 42834-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR