4-Bromo-3-methoxybenzyl alcohol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD71958-250MGEA 38 EUR
BLDPBD71958-250MG BLDPBD71958-1G
4-Bromo-3-methoxybenzyl alcohol 98%
4-Bromo-3-methoxybenzyl alcohol

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₉BrO₂
Molekulargewicht: 217,06 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11053671
CAS-Nummer: 17100-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR