2-Brom-5-phenyloxazol 95%

BLDPBD72010-1GEA 211 EUR
BLDPBD72010-1G BLDPBD72010-100MG BLDPBD72010-250MG
2-Brom-5-phenyloxazol 95%
2-Bromo-5-phenyloxazole

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₉H₆BrNO
Molekulargewicht: 224,0540 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD12828094
CAS-Nummer: 129053-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR