2-Chlor-4-methylbenzonitril 97%

BLDPBD73012-250MGEA 6 EUR
BLDPBD73012-250MG BLDPBD73012-25G BLDPBD73012-10G BLDPBD73012-100G BLDPBD73012-5G BLDPBD73012-1G
2-Chlor-4-methylbenzonitril 97%
2-Chlor-4-methylbenzonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆CIN
Molekulargewicht: 151,5900 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03094672
CAS-Nummer: 21423-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR