Methyl-2-chlor-5-iodbenzoat 98%

BLDPBD73198-5GEA 55 EUR
BLDPBD73198-5G BLDPBD73198-1G BLDPBD73198-250MG
Methyl-2-chlor-5-iodbenzoat 98%
Methyl-2-chlor-5-iodbenzoat

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₆ClIO₂
Molekulargewicht: 296,4900 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07779253
CAS-Nummer: 620621-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR