2-Chlorchinolin-3-boronsäure 96%

BLDPBD7344-1GEA 40 EUR
BLDPBD7344-1G BLDPBD7344-5G BLDPBD7344-250MG
2-Chlorchinolin-3-boronsäure 96%
2-Chlorchinolin-3-boronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₇BClNO₂
Molekulargewicht: 207,4214 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01114675
CAS-Nummer: 128676-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR