5-Methoxyisatoic anhydride 97%

BLDPBD7491-10GEA 270 EUR
BLDPBD7491-10G BLDPBD7491-5G BLDPBD7491-250MG BLDPBD7491-25G BLDPBD7491-1G
5-Methoxyisatoic anhydride 97%
5-Methoxyisatoic anhydride

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇NO₄
Molekulargewicht: 193,156 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02179411
CAS-Nummer: 37795-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR