2-Fluor-6-(trifluormethyl)benzylamin 96%

BLDPBD75036-25GEA 431 EUR
BLDPBD75036-25G BLDPBD75036-5G BLDPBD75036-250MG BLDPBD75036-1G BLDPBD75036-100MG
2-Fluor-6-(trifluormethyl)benzylamin 96%
2-Fluor-6-(trifluormethyl)benzylamin

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₇F₄N
Molekulargewicht: 193,14 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00061165
CAS-Nummer: 239087-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR