3-Amino-4-brom-5-phenyl-1H-pyrazol 95%

BLDPBD7523-1GEA 121 EUR
BLDPBD7523-1G BLDPBD7523-5G BLDPBD7523-250MG
3-Amino-4-brom-5-phenyl-1H-pyrazol 95%
3-Amino-4-bromo-5-phenyl-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₈BrN₃
Molekulargewicht: 238,09 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00082666
CAS-Nummer: 2845-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR