3-Iodo-5-methylpyridin-2-ol 98+%

BLDPBD75547-5GEA 288 EUR
BLDPBD75547-5G BLDPBD75547-1G BLDPBD75547-250MG
3-Iodo-5-methylpyridin-2-ol 98+%
3-Iodo-5-methylpyridin-2-ol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₆INO
Molekulargewicht: 235,03 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09037469
CAS-Nummer: 313678-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR