2-Hydroxy-3-(trifluormethyl)benzaldehyd 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD75596-5GEA 330 EUR
BLDPBD75596-5G BLDPBD75596-250MG BLDPBD75596-1G BLDPBD75596-100MG
2-Hydroxy-3-(trifluormethyl)benzaldehyd 98%
2-Hydroxy-3-(trifluoromethyl)benzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₅F₃O₂
Molekulargewicht: 190,12 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03788529
CAS-Nummer: 336628-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR