(R)-allylglycinol hydrochloride 95%

BLDPBD763906-250MGEA 212 EUR
BLDPBD763906-250MG BLDPBD763906-1G BLDPBD763906-100MG BLDPBD763906-50MG
(R)-allylglycinol hydrochloride 95%
(R)-allylglycinol hydrochloride

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₅H₁₁NO·ClH
Molekulargewicht: 137,608 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD11052925
CAS-Nummer: 926660-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR