(4-Chlorphenylthio)aceton 98%

BLDPBD7708-5GEA 42 EUR
BLDPBD7708-5G BLDPBD7708-25G BLDPBD7708-1G
(4-Chlorphenylthio)aceton 98%
(4-Chlorphenylthio)aceton

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉ClOS
Molekulargewicht: 200,6852 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013644
CAS-Nummer: 25784-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR