5-Amino-1-phenyl-1H-pyrazol 97%

BLDPBD7754-10GEA 92 EUR
BLDPBD7754-10G BLDPBD7754-5G BLDPBD7754-1G
5-Amino-1-phenyl-1H-pyrazol 97%
5-Amino-1-phenyl-1H-pyrazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₉N₃
Molekulargewicht: 159,1900 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00020734
CAS-Nummer: 826-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR