2-Nitro-4-pyridinecarboxylic acid 98%

BLDPBD77727-10GEA 374 EUR
BLDPBD77727-10G BLDPBD77727-250MG BLDPBD77727-25G BLDPBD77727-1G BLDPBD77727-5G
2-Nitro-4-pyridinecarboxylic acid 98%
2-Nitro-4-pyridinecarboxylic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₄N₂O₄
Molekulargewicht: 168,11 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD04114160
CAS-Nummer: 33225-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR