Methyl-2-methyl-5-nitrobenzoat 97%

BLDPBD7806-10GEA 18 EUR
BLDPBD7806-10G BLDPBD7806-25G BLDPBD7806-1G BLDPBD7806-100G BLDPBD7806-0.025G BLDPBD7806-5G
Methyl-2-methyl-5-nitrobenzoat 97%
Methyl-2-methyl-5-nitrobenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉NO₄
Molekulargewicht: 195,18 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00230865
CAS-Nummer: 77324-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR