Methyl-3-methyl-2-nitrobenzoat 97%

BLDPBD7807-10GEA 7 EUR
BLDPBD7807-10G BLDPBD7807-100G BLDPBD7807-25G BLDPBD7807-500G BLDPBD7807-5G
Methyl-3-methyl-2-nitrobenzoat 97%
Methyl-3-methyl-2-nitrobenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉NO₄
Molekulargewicht: 195,17 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01003947
CAS-Nummer: 5471-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR