2-(Pyridin-3-yl)aniline 95%

BLDPBD78462-5GEA 596 EUR
BLDPBD78462-5G BLDPBD78462-250MG BLDPBD78462-1G BLDPBD78462-100MG
2-(Pyridin-3-yl)aniline 95%
2-(Pyridin-3-yl)aniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₀N₂
Molekulargewicht: 170,21 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08060516
CAS-Nummer: 177202-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR