2-Chlor-5-(trifluormethoxy)phenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD8026-100MGEA 6 EUR
BLDPBD8026-100MG
2-Chlor-5-(trifluormethoxy)phenylboronsäure 98%
2-Chlor-5-(trifluormethoxy)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₅BClF₃O₃
Molekulargewicht: 240,37 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06656267
CAS-Nummer: 1022922-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR