3-(Trimethylsilylethinyl)pyridin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD81121-5GEA 72 EUR
BLDPBD81121-5G BLDPBD81121-1G BLDPBD81121-25G
3-(Trimethylsilylethinyl)pyridin 98%
3-(Trimethylsilylethinyl)pyridin

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₀H₁₃NSi
Molekulargewicht: 175,31 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD01863744
CAS-Nummer: 80673-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR