3,3'-Dimethyldiphenylamin 97%

BLDPBD81401-25GEA 338 EUR
BLDPBD81401-25G BLDPBD81401-5G BLDPBD81401-1G BLDPBD81401-250MG
3,3'-Dimethyldiphenylamin 97%
3,3'-Dimethyldiphenylamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₁₅N
Molekulargewicht: 197,28 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00059315
CAS-Nummer: 626-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR