3,5-Dichlorpyridazin 95%

BLDPBD82573-5GEA 87 EUR
BLDPBD82573-5G BLDPBD82573-10G BLDPBD82573-1G BLDPBD82573-100G BLDPBD82573-25G BLDPBD82573-250MG
3,5-Dichlorpyridazin 95%
3,5-Dichloropyridazine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₄H₂Cl₂N₂
Molekulargewicht: 148,9781 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10698048
CAS-Nummer: 1837-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR