4-Chlor-6,7-dimethoxychinazolin 98%

BLDPBD8279-1GEA 9 EUR
BLDPBD8279-1G BLDPBD8279-5G BLDPBD8279-10G BLDPBD8279-100G BLDPBD8279-25G
4-Chlor-6,7-dimethoxychinazolin 98%
4-Chloro-6,7-dimethoxyquinazoline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₀H₉ClN₂O₂
Molekulargewicht: 224,65 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01570172
CAS-Nummer: 13790-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR