3-Acetylbenzophenon 98%

BLDPBD83569-100MGEA 106 EUR
BLDPBD83569-100MG BLDPBD83569-1G BLDPBD83569-5G BLDPBD83569-250MG
3-Acetylbenzophenon 98%
3-Acetylbenzophenon

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₁₂O₂
Molekulargewicht: 224,2546 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01662280
CAS-Nummer: 66067-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR