3-Bromo-2-methylimidazo[1,2-a]pyridine 98%

BLDPBD84971-5GEA 396 EUR
BLDPBD84971-5G BLDPBD84971-1G BLDPBD84971-250MG BLDPBD84971-100MG
3-Bromo-2-methylimidazo[1,2-a]pyridine 98%
3-Bromo-2-methylimidazo[1,2-a]pyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₇BrN₂
Molekulargewicht: 211,059 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02251329
CAS-Nummer: 4805-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR