(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-4-[(2-methyl-2-propanyl)oxy]-4-oxobutansäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD8581-100GEA 58 EUR
BLDPBD8581-100G BLDPBD8581-10G BLDPBD8581-500G BLDPBD8581-25G
(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-4-[(2-methyl-2-propanyl)oxy]-4-oxobutansäure 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₂₃H₂₅NO₆
Molekulargewicht: 411,5 g/mol
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 71989-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR