3-Chlor-5-nitro-1H-indazol 97%

BLDPBD85960-10GEA 125 EUR
BLDPBD85960-10G BLDPBD85960-250MG BLDPBD85960-5G BLDPBD85960-1G
3-Chlor-5-nitro-1H-indazol 97%
3-Chloro-5-nitro-1H-indazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄ClN₃O₂
Molekulargewicht: 197,5800 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00005692
CAS-Nummer: 4812-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR