N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-(2-methyl-2-propanyl)-L-threonin 97%

BLDPBD8608-100GEA 88 EUR
BLDPBD8608-100G BLDPBD8608-10G BLDPBD8608-500G BLDPBD8608-25G BLDPBD8608-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-(2-methyl-2-propanyl)-L-threonin 97%
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₂₃H₂₇NO₅
Molekulargewicht: 397,5 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00077075
CAS-Nummer: 71989-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR