3-Chlorpropylacetat 98%

BLDPBD86195-10GEA 9 EUR
BLDPBD86195-0.025G BLDPBD86195-0.5G BLDPBD86195-10G BLDPBD86195-1000G BLDPBD86195-100G BLDPBD86195-5G BLDPBD86195-25G BLDPBD86195-500G
3-Chlorpropylacetat 98%
3-Chlorpropylacetat

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₉ClO₂
Molekulargewicht: 136,58 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00001000
CAS-Nummer: 628-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR