3'-Fluor-4'-(trifluormethyl)acetophenon 95%

BLDPBD86776-1GEA 26 EUR
BLDPBD86776-1G BLDPBD86776-5G BLDPBD86776-250MG
3'-Fluor-4'-(trifluormethyl)acetophenon 95%
3'-Fluor-4'-(trifluormethyl)acetophenon

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₆F₄O
Molekulargewicht: 206,14 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00236273
CAS-Nummer: 237761-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR