Isonipecotamid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD8718-10GEA 11 EUR
BLDPBD8718-10G BLDPBD8718-5G BLDPBD8718-100G BLDPBD8718-25G BLDPBD8718-500G
Isonipecotamid 95%
Isonipecotamid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₁₂N₂O
Molekulargewicht: 128,18 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00038012
CAS-Nummer: 39546-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR