3-Iod-4-methoxybenzaldehyd 98%

BLDPBD87469-1GEA 34 EUR
BLDPBD87469-1G BLDPBD87469-250MG BLDPBD87469-100MG BLDPBD87469-5G
3-Iod-4-methoxybenzaldehyd 98%
3-Iodo-4-methoxybenzaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₇IO₂
Molekulargewicht: 262,04 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01009500
CAS-Nummer: 2314-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR