3-Methoxybenzylhydrazine dihydrochloride 98+%

BLDPBD87771-1GEA 61 EUR
BLDPBD87771-1G BLDPBD87771-250MG BLDPBD87771-5G
3-Methoxybenzylhydrazine dihydrochloride 98+%
3-Methoxybenzylhydrazine dihydrochloride

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₂N₂O·2ClH
Molekulargewicht: 225,116 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06245507
CAS-Nummer: 849021-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR