3'-Methylacetanilid 98%

Synonyme: N-(3-Methylphenyl)acetamidEthansäure(3-methylphenyl)amid

BLDPBD88142-100GEA 27 EUR
BLDPBD88142-100G BLDPBD88142-25G
3'-Methylacetanilid 98%
3'-Methylacetanilid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₁NO
Molekulargewicht: 149,19 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00014962
CAS-Nummer: 537-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR