3-(Pyridin-3-yl)aniline 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD89070-1GEA 140 EUR
BLDPBD89070-1G BLDPBD89070-0.025G BLDPBD89070-100MG BLDPBD89070-250MG BLDPBD89070-25G BLDPBD89070-5G
3-(Pyridin-3-yl)aniline 95%
3-(Pyridin-3-yl)aniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₀N₂
Molekulargewicht: 170,211 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07368549
CAS-Nummer: 57976-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR