4-Methylnicotinamid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD89096-100MGEA 37 EUR
BLDPBD89096-100MG BLDPBD89096-250MG BLDPBD89096-1G
4-Methylnicotinamid 95%
4-Methylnicotinamid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₈N₂O
Molekulargewicht: 136,1512 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00234322
CAS-Nummer: 7250-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR