3-(Boc-amino)phenylboronsäurepinakolester 96%

Lieferant: BLD PHARMATECH GMBH

BLDPBD8923-5GEA 47 EUR
BLDPBD8923-5G BLDPBD8923-25G BLDPBD8923-10G BLDPBD8923-250MG BLDPBD8923-1G
3-(Boc-amino)phenylboronsäurepinakolester 96%
3-(Boc-amino)phenylboronsäurepinakolester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₇H₂₆BNO₄
Molekulargewicht: 319,21 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03789256
CAS-Nummer: 330793-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR