1-Boc-5-indolboronsäurepinakolester 98%

BLDPBD9048-5GEA 144 EUR
BLDPBD9048-5G BLDPBD9048-25G BLDPBD9048-1G
1-Boc-5-indolboronsäurepinakolester 98%
1-Boc-5-indolboronsäurepinakolester

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₉H₂₆BNO₄
Molekulargewicht: 343,2251 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD05663895
CAS-Nummer: 777061-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR