4-(Methansulfonylamino)benzonitril 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD91376-10GEA 225 EUR
BLDPBD91376-10G BLDPBD91376-1G BLDPBD91376-5G
4-(Methansulfonylamino)benzonitril 97%
4-(Methansulfonylamino)benzonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈N₂O₂S
Molekulargewicht: 196,23 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00204724
CAS-Nummer: 36268-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR