4-(1-Naphthyl)phenylboronsäure 97%

BLDPBD91481-25GEA 104 EUR
BLDPBD91481-25G BLDPBD91481-1G BLDPBD91481-100G BLDPBD91481-5G BLDPBD91481-10G
4-(1-Naphthyl)phenylboronsäure 97%
4-(1-Naphthyl)phenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₁₃BO₂
Molekulargewicht: 248,09 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08669639
CAS-Nummer: 870774-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR