4-Amino-3-nitrobenzolsulfonamid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD93847-25GEA 1153 EUR
BLDPBD93847-25G BLDPBD93847-100MG BLDPBD93847-1G BLDPBD93847-250MG BLDPBD93847-5G
4-Amino-3-nitrobenzolsulfonamid 95%
4-Amino-3-nitrobenzolsulfonamid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₇N₃O₄S
Molekulargewicht: 217,2000 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00116612
CAS-Nummer: 2360-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR