4-(Aminomethyl)tetrahydropyran HCl 98%
|
|
BLDPBD94171-1GEA
17
EUR
BLDPBD94171-1G
BLDPBD94171-5G
BLDPBD94171-25G
4-(Aminomethyl)tetrahydropyran HCl 98%
4-(Aminomethyl)tetrahydropyran HCl
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₆H₁₄ClNO Molekulargewicht: 151,6300 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03411928 CAS-Nummer: 389621-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...