4-Brom-2,5-dimethoxybenzaldehyd 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD94769-1GEA 10 EUR
BLDPBD94769-1G BLDPBD94769-100G BLDPBD94769-10G BLDPBD94769-5G BLDPBD94769-250MG BLDPBD94769-25G
4-Brom-2,5-dimethoxybenzaldehyd 95%
4-Bromo-2,5-dimethoxybenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉BrO₃
Molekulargewicht: 245,08 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00156964
CAS-Nummer: 31558-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR