4'-Brom-2'-nitroacetanilid 97%

BLDPBD94930-5GEA 98 EUR
BLDPBD94930-5G BLDPBD94930-250MG BLDPBD94930-1G BLDPBD94930-25G
4'-Brom-2'-nitroacetanilid 97%
4'-Brom-2'-nitroacetanilid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇BrN₂O₃
Molekulargewicht: 259,0600 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00024299
CAS-Nummer: 881-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR