4-Hydroxy-3-(trifluormethyl)benzonitril 98%

BLDPBD97667-5GEA 79 EUR
BLDPBD97667-5G BLDPBD97667-1G BLDPBD97667-25G BLDPBD97667-10G BLDPBD97667-250MG
4-Hydroxy-3-(trifluormethyl)benzonitril 98%
4-Hydroxy-3-(trifluormethyl)benzonitril

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₄F₃NO
Molekulargewicht: 187,12 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06797882
CAS-Nummer: 124811-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR